missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAGC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RRAGC |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
16885305
|
Proteintech
26989-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16875305
|
Proteintech
26989-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This gene encodes a protein that is required for the formation of hypusine, a unique amino acid formed by the posttranslational modification of only one protein, eukaryotic translation initiation factor 5A. The encoded protein catalyzes the first step in hypusine formation by transferring the butylamine moiety of spermidine to a specific lysine residue of the eukaryotic translation initiation factor 5A precursor, forming an intermediate deoxyhypusine residue. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Spezifikation
| RRAGC | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q99K70, Q9HB90 | |
| 54170, 64121 | |
| RRAGC Fusion Protein Ag25780 | |
| Primary | |
| -20°C | |
| RRAGC |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GTPase interacting protein 2, GTR2, Rag C, RAGC, Ras related GTP binding C, RRAGC, TIB929 | |
| Rragc | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts