missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRAGC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584256
This item is not returnable.
View return policy
Description
Immunogen sequence: MSLQYGAEET PLAGSYGAAD SFPKDFGYGV EEEEEE Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
RRAGC (Ras-related GTP-binding protein C) is a monomeric guanine nucleotide-binding protein that plays a crucial role in the cellular response to amino acid availability through regulations of the mTORC1 signaling cascade. RRAGC forms a heterodimer with RRAGA and RRAGB and is primarily localized to the cytoplasm. It cycles between an inactive GTP-bound and an active GDP-bound form. RRAGC, in its active form, participates in the re-localization of mTORC1 to the lysosomes and its subsequent activation by the GTPase RHEB. This re-localization is a crucial step in the activation of the TOR signaling cascade by amino acids.
Specifications
| RRAGC | |
| Polyclonal | |
| Unconjugated | |
| Rragc | |
| AU041672; GTPase-interacting protein 2; GTR2; Rag C; Rag C protein; RAGC; Ras related GTP binding C; Ras-related GTP binding C; ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929; YGR163W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HB90 | |
| Rragc | |
| Recombinant protein corresponding to Human RRAGC. Recombinant protein control fragment (Product #RP-106640). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction