missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RRAGC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29967
This item is not returnable.
View return policy
Description
RRAGC Polyclonal antibody specifically detects RRAGC in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| RRAGC | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FLJ13311, GTPase-interacting protein 2, GTR2, Rag C, Rag C protein, RAGC, Ras-related GTP binding C, ras-related GTP-binding protein C, TIB929 | |
| Between 1 and 50 | |
| 100 μg | |
| Apoptosis, mTOR Pathway | |
| 64121 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction