missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RQ 00203078
GREENER_CHOICE

Product Code. p-5545595
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18778218

Brand: Tocris Bioscience™ 5388/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TRPM8 antagonist; orally active

RQ 00203078 is a potent TRPM8 antagonist (IC50 values are 5.3 and 8.3 nM for rat and human channels respectively). Exhibits >350-fold selectivity for TRPM8 over TRPV4, TRPV1 and TRPA1. Reduces HSC3 and HSC4 oral squamous carcinoma cell migration and invasion in vitro. Also attenuates icilin -induced wet dog shakes in rats. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type RQ 00203078
Purity >98%
Molecular Weight (g/mol) 554.85

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.