missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPUSD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532069
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse brain. Predicted reactivity: Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNA Pseudouridine Synthase D3 (RPUSD3) is a gene that encodes a pseudouridine synthase, an enzyme responsible for catalyzing the isomerization of uridine to pseudouridine in RNA molecules. This post-transcriptional modification is crucial for the stability and function of tRNAs and rRNAs. RPUSD3, along with other pseudouridine synthases such as PUS1, PUSL1, and TRUB2, is predicted to localize to mitochondria, indicating its role in mitochondrial RNA processing and maturation. The pseudouridylation activity provided by RPUSD3 is essential for the proper folding and functionality of mitochondrial RNAs, which are fundamental for the biogenesis of the mitochondrial electron transport chain and ATP synthase. While detailed structural insights into RPUSD3 are not yet fully characterized, its enzymatic function is critical in maintaining cellular energy metabolism and overall mitochondrial health.
Specifications
| RPUSD3 | |
| Polyclonal | |
| Unconjugated | |
| RPUSD3 | |
| AI527266; Mitochondrial mRNA pseudouridine synthase RPUSD3; RGD1307582; RNA pseudouridylate synthase domain containing 3; RNA pseudouridylate synthase domain-containing protein 3; Rpusd3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 101122, 285367 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.65 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14AI6, Q6P087 | |
| RPUSD3 | |
| Recombinant protein encompassing a sequence within the center region of human RPUSD3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction