Learn More
RPUSD2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25707-1-AP-20UL
This item is not returnable.
View return policy
Description
The RPUSD2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RPUSD2. This antibody recognizes human antigen. The RPUSD2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| RPUSD2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 27079 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human |