missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RPTOR Polyclonal antibody specifically detects RPTOR in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RPTOR |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 1% BSA, 20% Glycerol (pH7) |
| Gene Alias | KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide corresponding to residues around human RPTOR (phospho Ser792). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?