missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPSA Recombinant Rabbit Monoclonal Antibody (K01_1M23)

Product Code. 30829916
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829916 100 μL
30826727 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829916 Supplier Invitrogen™ Supplier No. MA563299
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RPSA Recombinant Monoclonal Antibody for Western Blot

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Many of the effects of laminin are mediated through interactions with cell surface receptors. These receptors include members of the integrin family, as well as non-integrin laminin-binding proteins. This gene encodes a high-affinity, non-integrin family, laminin receptor 1. This receptor has been variously called 67 kD laminin receptor, 37 kD laminin receptor precursor (37LRP) and p40 ribosome-associated protein. The amino acid sequence of laminin receptor 1 is highly conserved through evolution, suggesting a key biological function. It has been observed that the level of the laminin receptor transcript is higher in colon carcinoma tissue and lung cancer cell line than their normal counterparts. Also, there is a correlation between the upregulation of this polypeptide in cancer cells and their invasive and metastatic phenotype. Multiple copies of this gene exist, however, most of them are pseudogenes thought to have arisen from retropositional events. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPSA
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1M23
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene RPSA
Gene Accession No. P08865, P14206, P38983
Gene Alias 37 kDa laminin receptor; 37 kDa laminin receptor precursor; 37 kDa laminin receptor precursor {ECO:0000255; 37 kDa oncofetal antigen; 37/67 kDa laminin receptor; 37/67 kDa laminin receptor {ECO:0000255; 37LRP; 37LRP {ECO:0000255; 40S ribosomal protein SA; 40S ribosomal protein SA {ECO:0000255; 67 kDa laminin receptor; 67 kDa laminin receptor {ECO:0000255; 67kDa; 67LR; 67LR {ECO:0000255; AL022858; colon carcinoma laminin-binding protein; HAMAP-Rule:MF_03016}; ICAS; LAMBR; Laminin receptor 1; laminin receptor 1 (67kD, ribosomal protein SA); laminin receptor 1 (ribosomal protein SA); laminin receptor 1 {ECO:0000255; laminin-binding protein precursor p40; laminin-binding protein precursor p40 {ECO:0000255; LamR; lamR {ECO:0000255; LAMR1; Lamrl1; LBP; LBP/p40; LBP/p40 {ECO:0000255; LRP; LRP/LR; LRP/LR {ECO:0000255; MLR; Multidrug resistance-associated protein MGr1-Ag; NEM/1CHD4; OFA/iLRP; p40; P40-3; P40-3, functional; P40-8; P40-8, functional; ribosomal protein SA; Rpsa; rssa; SA; Small ribosomal subunit protein uS2
Gene Symbols RPSA
Host Species Rabbit
Immunogen A synthetic peptide of human 67kDa Laminin Receptor.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16785, 29236, 3921
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.