missing translation for 'onlineSavingsMsg'
Learn More

RPSA Antibody (RPSA/6332R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18331556 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331556 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331556 Supplier Novus Biologicals Supplier No. NBP314110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RPSA Monoclonal antibody specifically detects RPSA in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPSA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RPSA/6332R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Formulation 10mM PBS without Preservative
Gene Alias 37 kDa laminin receptor precursor, 37/67 kDa laminin receptor, 37LRPNEM/1CHD4, 67 kDa laminin receptor, 67kD, ribosomal protein SA, 67LR, Colon carcinoma laminin-binding protein, LAMBR37 kDa laminin receptor, laminin binding protein, Laminin receptor 1, laminin receptor 1 (67kD, ribosomal protein SA), Laminin-binding protein precursor p40, lamR, LAMR 1, LAMR140S ribosomal protein SA, LBP, LBP/p40, LRP, LRP/LR, Multidrug resistance-associated protein MGr1-Ag, p40, ribosomal protein SA
Host Species Rabbit
Immunogen Recombinant full-length human RPSA protein (Uniprot: P08865)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Core ESC Like Genes, Extracellular Matrix, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3921
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.