missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS8 Recombinant Rabbit Monoclonal Antibody (24GB4120)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557227
This item is not returnable.
View return policy
Description
RPS8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal tumors and colon polyps compared to matched normal colonic mucosa has been observed. This gene is co-transcribed with the small nucleolar RNA genes U38A, U38B, U39, and U40, which are located in its fourth, fifth, first, and second introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS8 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Rps8 | |
| 40S ribosomal protein S8; OK/SW-cl.83; ribosomal protein S8; Rps8; S8; Small ribosomal subunit protein eS8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20116, 6202, 65136 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4120 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62241, P62242, P62243 | |
| Rps8 | |
| A synthesized peptide derived from human RPS8 (aa 1-208). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction