missing translation for 'onlineSavingsMsg'
Learn More

RPS6KC1 Antibody (OTI10F11), DyLight 594, Novus Biologicals™

Product Code. 18400787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400787 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400787 Supplier Novus Biologicals Supplier No. NBP273946DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RPS6KC1 Monoclonal antibody specifically detects RPS6KC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS6KC1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI10F11
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11.1, humS6PKh1, ribosomal protein S6 kinase delta-1, ribosomal protein S6 kinase, 52kD, polypeptide 1,52 kDa ribosomal protein S6 kinase, ribosomal protein S6 kinase, 52kDa, polypeptide 1, Ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein, RPK118, S6K-delta-1, SPHK1-binding protein
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-305 of human RPS6KC1 (NP_036556) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 26750
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.