missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94674-0.1ml
This item is not returnable.
View return policy
Description
RPS6KC1 Polyclonal antibody specifically detects RPS6KC1 in Human samples. It is validated for Western Blot
Specifications
| RPS6KC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 2.7.11.1, humS6PKh1, ribosomal protein S6 kinase delta-1, ribosomal protein S6 kinase, 52kD, polypeptide 1,52 kDa ribosomal protein S6 kinase, ribosomal protein S6 kinase, 52kDa, polypeptide 1, Ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein, RPK118, S6K-delta-1, SPHK1-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human RPS6KC1 (NP_036556.2). DSSALGAVASDSEQSKTEEERESRSLFPGSLKPKLGKRDYLEKAGELIKLALKKEEEDDYEAASDFYRKGVDLLLEGVQGESSPTRREAVKRRTAEYLMRAESISSLYGKPQLDDVSQPPGSLSSRPLWNLRSPAEELKAFRVLGVIDKVLLVMDTRTEQTFILKGLRKSSEYSRNRKTIIPRCVPNMVCLHKYIISEESV | |
| 0.1 mL | |
| Protein Kinase | |
| 26750 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction