missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS6KB2 Recombinant Rabbit Monoclonal Antibody (24GB1235)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557692
This item is not returnable.
View return policy
Description
RPS6KB2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
| RPS6KB2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Rps6kb2 | |
| 70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; RPS6KB2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14B; SRK; STK14B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361696, 58988, 6199 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1235 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBS0, Q9Z1M4 | |
| Rps6kb2 | |
| A synthesized peptide derived from human P70 S6 Kinase beta (aa 1-120). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction