missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPS6KA6 (RSK4), GST Tag Recombinant Protein

Product Code. 15408663
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15408663

Brand: Invitrogen™ PR8170A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains. RSK4 encodes a member of ribosomal S6 kinase family, serine-threonine protein kinases which are regulated by growth factors. The encoded protein may be distinct from other members of this family, however, as studies suggest it is not growth factor dependent and may not participate in the same signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UK32
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 27330
Molecular Weight (g/mol) 111.3 kDa
Name Human RPS6KA6 (RSK4), GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias 2610524K04Rik; 90 kDa ribosomal protein S6 kinase 6; p90-RSK 6; p90RSK6; pp90RSK4; RGD1560817; ribosomal protein S6 kinase A6; ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90 kDa, polypeptide 6; ribosomal S6 kinase 4; Rps6ka6; RSK4; RSK-4; S6KA6; S6K-alpha 6; S6K-alpha-6
Common Name RSK4
KinaseFamily RSK (RPS6K) Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.