missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KA6, Mouse anti-Human, Clone: 8D2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00027330-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: RIGNGKFSLSGGNWDNISDGAKDLLSHMLHMDPHQRYTAEQILKHSWITHRDQLPNDQPKRNDVSHVVKGAMVATYSALTHKTFQPVLEPVAASSLAQRRSMKKRTSTGLSpecifications
| RPS6KA6 | |
| Monoclonal | |
| Unconjugated | |
| ribosomal protein S6 kinase, 90kDa, polypeptide 6 | |
| RSK4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 8D2 | |
| In 1x PBS, pH 7.4 | |
| NM_014496 | |
| RPS6KA6 | |
| RPS6KA6 (NP_055311.1, 636 a.a. ∼ 745 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 27330 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction