missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPS6KA3 (RSK2), His Tag Recombinant Protein

Product Code. 11513437
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11513437

Brand: Invitrogen™ PV3323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

RPS6KA3 (RSK2) is a serine/threonine kinase that modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation. RPS6KA3 contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Mutations in the gene have been associated with Coffin-Lowry syndrome (CLS).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51812
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 2mM DTT, 150mM NaCl, 0.05mM EDTA, 50% glycerol, 0.05% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 6197
Molecular Weight (g/mol) 86.3 kDa
Name Human RPS6KA3 (RSK2), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 90 kDa ribosomal protein S6 kinase 3; C130006E23; CLS; HU-3; Insulin-stimulated protein kinase 1; ISPK1; ISPK-1; MAP kinase-activated protein kinase 1b; MAPK-activated protein kinase 1b; MAPKAP kinase 1b; MAPKAPK1B; MAPKAPK-1b; MPK-9; MRX19; p90-RSK 3; p90-RSK2; p90RSK3; pp90RSK2; RGD1563860; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase A3; ribosomal protein S6 kinase alpha-3; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal S6 kinase 2; Rps6ka3; Rps6ka-rs1; RSK; Rsk2; RSK-2; S6K-alpha3; S6K-alpha-3
Common Name RSK2
Gene Symbol RPS6KA3
KinaseFamily RSK (RPS6K) Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: AGC Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry RPS6KA3
Target Gene ribosomal protein S6 kinase, 90 kDa, polypeptide 3
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.