missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KA2, Mouse anti-Human, Clone: 1F6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006196-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: YALSGGNWDSISDAAKDVVSKMLHVDPHQRLTAMQVLKHPWVVNREYLSPNQLSRQDVHLVKGAMAATYFALNRTPQAPRLEPVLSSNLAQRRGMKRLTSTRLSpecifications
| RPS6KA2 | |
| Monoclonal | |
| Unconjugated | |
| ribosomal protein S6 kinase, 90kDa, polypeptide 2 | |
| HU-2/MAPKAPK1C/RSK/RSK3/S6K-alpha/S6K-alpha2/p90-RSK3/pp90RSK3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), In situ PLA, KnockDown, Western Blot | |
| 1F6 | |
| In 1x PBS, pH 7.4 | |
| BC002363 | |
| RPS6KA2 | |
| RPS6KA2 (AAH02363, 631 a.a. ∼ 733 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6196 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction