missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS4X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14799-1-AP-20UL
This item is not returnable.
View return policy
Description
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.Specifications
| RPS4X | |
| Polyclonal | |
| Unconjugated | |
| RPS4X | |
| CCG2, DXS306, ribosomal protein S4, X linked, RPS4, RPS4X, SCAR, SCR10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100362640, 20102, 6191 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62701, P62702, P62703 | |
| RPS4X | |
| RPS4X Fusion Protein Ag6513 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction