missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS4X Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551051
This item is not returnable.
View return policy
Description
RPS4X Polyclonal Antibody detects endogenous levels of RPS4X protein.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.
Specifications
| RPS4X | |
| Polyclonal | |
| Unconjugated | |
| RPS4X | |
| 40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100362640, 20102, 6191 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62701, P62702, P62703 | |
| RPS4X | |
| Synthesized peptide derived from the Internal region of human Ribosomal Protein S4X. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction