missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS4X Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82761
This item is not returnable.
View return policy
Description
RPS4X Polyclonal specifically detects RPS4X in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RPS4X | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CCG2Single copy abundant mRNA protein, DXS306, FLJ40595, ribosomal protein S4, X-linked, RPS4cell cycle gene 2, SCAR40S ribosomal protein S4, X isoform, SCR10ribosomal protein S4X isoform, single-copy abundant mRNA | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RPS4X | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KVRKITVGVKGIPHLVTHDARTIRYPDPVIKVNDTVQIDLGTGKIINFIKFDTGNLCMVIGGANLGRVGVITNRERHPGSFDVVHVKDANGNSFATRLSNIFVIGNGNKPWISLPRGKGIRLTVA | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6191 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction