missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145009
This item is not returnable.
View return policy
Description
Immunogen sequence: QLIKNGKKIT AFVPNDGCLN FIEENDEVLV AGFGRKGHAV GDIPGVRFKV VKVANVSLLA LYKGKKERPR S Antigen sequence identity: Mouse 100%; Rat 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S12P family of ribosomal proteins. It is located in the cytoplasm. The protein shares significant amino acid similarity with S. cerevisiae ribosomal protein S28. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq, Jul 2008].
Specifications
| RPS23 | |
| Polyclonal | |
| Unconjugated | |
| RPS23 | |
| 2410044J15Rik; 40S ribosomal protein S23; homolog of yeast ribosomal protein S28; ribosomal protein S23; RPS23; S23; Small ribosomal subunit protein uS12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62266 | |
| RPS23 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 73-143. Recombinant protein control fragment (Product #RP-109799). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction