missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS23 Antibody (YWHAE/8636R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23780-20ug
This item is not returnable.
View return policy
Description
RPS23 Monoclonal antibody specifically detects RPS23 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RPS23 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| FLJ35016, homolog of yeast ribosomal protein S28, ribosomal protein S23,40S ribosomal protein S23 | |
| Recombinant human full-length RPS23 protein | |
| 20 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6228 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| YWHAE/8636R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction