missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS21 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £290.00
Specifications
| Antigen | RPS21 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18343804
|
Novus Biologicals
NBP3-15571-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392026
|
Novus Biologicals
NBP3-15571-100UL |
100 μg |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPS21 Polyclonal antibody specifically detects RPS21 in Human, Mouse samples. It is validated for Western BlotSpecifications
| RPS21 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 6227 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| ribosomal protein S21,40S ribosomal protein S21,8.2 kDa differentiation factor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-81 of human RPS21 (NP_001015.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title