missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116866
This item is not returnable.
View return policy
Description
Positive Samples: LO2, K-562, Mouse pancreas, Mouse liver Immunogen sequence: MQNDAGEFVD LYVPRKCSAS NRIIGAKDHA SIQMNVAEVD KVTGRFNGQF KTYAICGAIR RMGESDDSIL RLAKADGIVS K.
Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
Specifications
| RPS21 | |
| Polyclonal | |
| Unconjugated | |
| RPS21 | |
| 1810049N11Rik; 2410030A14Rik; 40S ribosomal protein S21; 8.2 kDa differentiation factor; HLDF; human leukemia differentiation factor; LOC615178 protein; ribosomal protein S21; rps21; S21; Small ribosomal subunit protein eS21; zgc:56642; zgc:86816 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6227, 66481 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.54 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P63220, Q9CQR2 | |
| RPS21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-81 of human RPS21 (NP_0010151). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction