Learn More
Invitrogen™ RPS20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120087
Description
Positive test controls include: HeLa, HT-1080, Mouse kidney, Mouse liver, Rat liver. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MAFKDTGKTP VEPEVAIHRI RITLTSRNVK SLEKVCADLI RGAKEKNLKV KGPVRMPTKT LRITTRKTPC GEGSKTWDRF QMRIHKRLID LHSPSEIVKQ ITSISIEPGV EVEVTIADA.
Mammalian ribosomal proteins are encoded by multigene families that consist of processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein S20, also known as RPS20, is a 119 amino acid cytoplasmic protein that is a component of the 40S ribosomal subunit. Co-transcribed with the small nucleolar RNA gene U54, Ribosomal Protein S20 is a primary binding protein (it binds independently to its target protein) that interacts with both the 5' and 3' minor domains of 16S ribosomal RNA (rRNA). Through its interactions with 16S rRNA, Ribosomal Protein S20 is thought to play a key role in nucleating the assembly of the 30S ribosomal subunit. Like most ribosomal protein-coding genes, the gene encoding Ribosomal Protein S20 is dispersed throughout the genome and exists as multiple processed pseudogenes.
Specifications
| RPS20 | |
| Polyclonal | |
| Unconjugated | |
| RPS20 | |
| 40S ribosomal protein S20; 4632426K06Rik; AFAR; AFAR1; AKR7; Dsk4; ribosomal protein S20; Rps20; S20; Small ribosomal subunit protein uS10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 122772, 6224, 67427 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P60866, P60867, P60868 | |
| RPS20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-119 of human RPS20 (NP_001014.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.