missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RPS20 Antibody (SR3012), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27729-100ul
This item is not returnable.
View return policy
Description
RPS20 Monoclonal antibody specifically detects RPS20 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| RPS20 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3012 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| 40S ribosomal protein S20, FLJ27451, MGC102930, ribosomal protein S20 | |
| A synthesized peptide derived from human RPS20 (Uniprot #: P60866) | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 6224 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction