missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£231.00 - £470.00
Specifications
| Antigen | RPS20 |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18466920
|
Novus Biologicals
NBP1-80804-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257935
|
Novus Biologicals
NBP1-80804 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPS20 Polyclonal specifically detects RPS20 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| RPS20 | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 40S ribosomal protein S20, FLJ27451, MGC102930, ribosomal protein S20 | |
| RPS20 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.1mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P60866 | |
| 6224 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MAFKDTGKTPVEPEVAIHRIRITLTSRNVKSLEKVCADLIRGAKEKNLKVKGPVRMPTKTLRITTRKTPCGEGSKTWDRFQMRIHKRLIDLHSPSEIVKQITSISIEPGVEVEVTIA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 13 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title