missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14852F
This item is not returnable.
View return policy
Description
RPS2 Polyclonal antibody specifically detects RPS2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| RPS2 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| LLREP3, MGC102851, MGC117344, MGC117345,40S ribosomal protein S4, OK/KNS-cl.6,40S ribosomal protein S2, Protein LLRep3, ribosomal protein S2, RPS4 | |
| This antibody maps to a region between residue 50 and 100 of human Ribosomal Protein S2 using the numbering given in entry NP_002943.2 (GeneID 6187). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6187 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction