missing translation for 'onlineSavingsMsg'
Learn More

RPS2 Antibody (3F5), Novus Biologicals™

Product Code. 18322889 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322889 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322889 Supplier Novus Biologicals Supplier No. H00006187M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RPS2 Monoclonal antibody specifically detects RPS2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3F5
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002943.2
Gene Alias LLREP3, MGC102851, MGC117344, MGC117345,40S ribosomal protein S4, OK/KNS-cl.6,40S ribosomal protein S2, Protein LLRep3, ribosomal protein S2, RPS4
Host Species Mouse
Immunogen RPS2 (NP_002943.2, 47 a.a. ~ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GRGARGGKAEDKEWMPVTKLGRLVKDMKIKSLEEIYLFSLPIKESEIIDFFLGASLKDEVLKIMPVQKQTRAGQRTRFKAFVAIGDYNGHVGLGVKCSKEVATAIRGAIILAKLSIVPVRRGYW
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6187
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.