missing translation for 'onlineSavingsMsg'
Learn More

RPS19 Rabbit anti-Human, Mouse, Rat, Clone: 4V3S7, Novus Biologicals™

Product Code. 18319441
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319441 20 μg 20µL
18344952 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18319441 Supplier Bio-Techne Supplier No. NBP31624420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RPS19 Monoclonal antibody specifically detects RPS19 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS19
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 4V3S7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DBADBA1,40S ribosomal protein S19, ribosomal protein S19
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 66-145 of human RPS19 (P39019).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6223
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.