missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS19 Antibody (7E6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | RPS19 |
|---|---|
| Clone | 7E6 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032616
|
Novus Biologicals
NBP3-26339-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30024602
|
Novus Biologicals
NBP3-26339-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPS19 Monoclonal antibody specifically detects RPS19 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
| RPS19 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DBADBA1,40S ribosomal protein S19, ribosomal protein S19 | |
| A synthesized peptide derived from Human RPS19 [UniProt P39019] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7E6 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 6223 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title