missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565916
This item is not returnable.
View return policy
Description
Immunogen sequence: QKDVKDGKYSQ VLANGLDNKL REDLERLKKI RAHRGL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S13P family of ribosomal proteins. It is located in the cytoplasm. The gene product of the E. coli ortholog (ribosomal protein S13) is involved in the binding of fMet-tRNA, and thus, in the initiation of translation. This gene is an ortholog of mouse Ke3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS18 | |
| Polyclonal | |
| Unconjugated | |
| RPS18 | |
| 40S ribosomal protein S18; D6S218E; H-2Ke3; H2-Ke3; HKE3; KE3; KE-3; rhabdomyosarcoma antigen MU-RMS-40.21; ribosomal protein S18; RPS18; S18; Small ribosomal subunit protein uS13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6222 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62269 | |
| RPS18 | |
| Recombinant Human RPS18. Recombinant protein control fragment (Product #RP-106593). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion