missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS17 Polyclonal Antibody

RPS17 Polyclonal Antibody
Brand: Invitrogen PA575600
This item is not returnable.
View return policy
Description
RPS17 Polyclonal Antibody for Western Blot
Specifications
| RPS17 | |
| Polyclonal | |
| Unconjugated | |
| Rps17 | |
| 40S ribosomal protein S17; DBA4; ribosomal protein S17; Rps17; RPS17L; RPS17L1; RPS17L2; S17; Small ribosomal subunit protein eS17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20068, 29286, 6218 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| P04644, P08708, P63276 | |
| Rps17 | |
| Synthetic peptide corresponding to residues in Human RPS17. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction