Learn More
RPS16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15603-1-AP-150UL
 View more versions of this product
Product Code. 16864844
  • £349.00 / 150µL
Description
15603-1-AP is a rabbit polyclonal antibody raised against full-length RPS16 of human origin.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S9P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
Specifications
RPS16
Polyclonal
Unconjugated
RPS16
40S ribosomal protein S16, ribosomal protein S16, RPS16
Rabbit
Antigen Affinity Chromatography
RUO
140655, 20055, 6217
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14131, P62249, P62250
RPS16
RPS16 Fusion Protein Ag7986
150 μL
Primary
Human, Mouse, Rat