missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563486

Product Code. 15741262

  • £475.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SMVGVYNGKT FNQVEIKPEM IGHYLGEFSI TYKPVKHGRP GIGATHSSRF IPLK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19P family of ribosomal proteins. It is located in the cytoplasm. This gene has been found to be activated in various tumors, such as insulinomas, esophageal cancers, and colon cancers. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPS15
Polyclonal
Unconjugated
RPS15
40S ribosomal protein S15; homolog of rat insulinoma; insulinoma; insulinoma protein; ribosomal protein S15; rig; RIG protein; RPS15; S15; Small ribosomal subunit protein uS19
Rabbit
Antigen affinity chromatography
RUO
6209
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P62841
RPS15
Recombinant protein corresponding to Human RPS15. Recombinant protein control fragment (Product #RP-101215).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.