missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA527546

Product Code. 13411657

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, MCF-7, MDA-MB-231. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (98%), Xenopus laevis (95%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19P family of ribosomal proteins. It is located in the cytoplasm. This gene has been found to be activated in various tumors, such as insulinomas, esophageal cancers, and colon cancers. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPS15
Polyclonal
Unconjugated
RPS15
40S ribosomal protein S15; homolog of rat insulinoma; insulinoma; insulinoma protein; ribosomal protein S15; rig; RIG protein; RPS15; S15; Small ribosomal subunit protein uS19
Rabbit
Antigen affinity chromatography
RUO
20054, 6209
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
P62841, P62843
RPS15
Recombinant protein encompassing a sequence within the center region of human RPS15. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.