missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106433
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RPS14.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS14 is a component of the 40S subunit. The protein belongs to the S11P family of ribosomal proteins. It is located in the cytoplasm. Transcript variants utilizing alternative transcription initiation sites have been described in the literature. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S11P family of ribosomal proteins. It is located in the cytoplasm. Transcript variants utilizing alternative transcription initiation sites have been described in the literature. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. In Chinese hamster ovary cells, mutations in this gene can lead to resistance to emetine, a protein synthesis inhibitor. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| RPS14 | |
| Polyclonal | |
| Unconjugated | |
| RPS14 | |
| 2600014J02Rik; 40S ribosomal protein S14; AL023078; emetine resistance; EMTB; fa92e08; PRO2640; ribosomal protein S14; Rps14; S14; similar to ribosomal protein S14; Small ribosomal subunit protein uS11; Unknown (protein for MGC:127734); wu:fa92e08; zgc:73215 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20044, 29284, 6208 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13471, P62263, P62264 | |
| RPS14 | |
| A synthesized peptide derived from human RPS14(Accession P62263), corresponding to amino acid residues D80-E130. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction