missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS14 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22319CL1
This item is not returnable.
View return policy
Description
RPS14 Polyclonal antibody specifically detects RPS14 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| RPS14 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| emetine resistance, EMTB40S ribosomal protein S14, ribosomal protein S14 | |
| The immunogen this antibody was made to, maps to a region between residue 101 to 151 of human Ribosomal Protein S14 using the numbering given in entry NP_001020242.1 (GeneID 6208). | |
| 0.1 mL | |
| metabolism | |
| 6208 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction