missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104281
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Ribosomal Protein S13.
RPS13, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S15P family of ribosomal proteins. It is located in the cytoplasm. The protein has been shown to bind to the 5.8S rRNA in rat. The gene product of the E. coli ortholog (ribosomal protein S15) functions at early steps in ribosome assembly. This gene is co-transcribed with two U14 small nucleolar RNA genes, which are located in its third and fifth introns. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS13 | |
| Polyclonal | |
| Unconjugated | |
| Rps13 | |
| 2700063M04Rik; 40S ribosomal protein S13; ribosomal protein S13; Rps13; S13; Small ribosomal subunit protein uS15 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 161477, 6207, 68052 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62277, P62278, P62301 | |
| Rps13 | |
| A synthesized peptide derived from human RPS13(Accession P62277), corresponding to amino acid residues K78-Y128. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction