missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16490-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes the 15 kDa RPS12 protein.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S12E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal cancers compared to matched normal colonic mucosa has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
| RPS12 | |
| Polyclonal | |
| Unconjugated | |
| RPS12 | |
| 40S ribosomal protein S12, ribosomal protein S12, RPS12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20042, 6206, 65139 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P25398, P63323, P63324 | |
| RPS12 | |
| RPS12 Fusion Protein Ag9615 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction