missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101368
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RPS11.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S17P family of ribosomal proteins. It is located in the cytoplasm. The gene product of the E. coli ortholog (ribosomal protein S17) is thought to be involved in the recognition of termination codons. This gene is co-transcribed with a small nucleolar RNA gene, which is located in its third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS11 | |
| Polyclonal | |
| Unconjugated | |
| RPS11 | |
| 40S ribosomal protein S11; ribosomal protein S11; RPS 11; RPS11; S11; Small ribosomal subunit protein uS17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27207, 6205, 81774 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62280, P62281, P62282 | |
| RPS11 | |
| A synthesized peptide derived from human RPS11, corresponding to a region within C-terminal amino acids. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction