missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS11 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22288CL1
This item is not returnable.
View return policy
Description
RPS11 Polyclonal antibody specifically detects RPS11 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| RPS11 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6205 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| ribosomal protein S11,40S ribosomal protein S11 | |
| The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Ribosomal Protein S11 using the numbering given in entry NP_001006.1 (GeneID 6205). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction