missing translation for 'onlineSavingsMsg'
Learn More

RPRD2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18423607 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423607 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423607 Supplier Novus Biologicals Supplier No. NBP197932AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

RPRD2 Polyclonal antibody specifically detects RPRD2 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPRD2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias HSPC099, KIAA0460FLJ32145, regulation of nuclear pre-mRNA domain containing 2, regulation of nuclear pre-mRNA domain-containing protein 2
Host Species Goat
Immunogen The immunogen this antibody was made to, maps to a region between residues 1000 and 1050 of human RPRD2 using the numbering given in TrEMBL entry Q5VT52 (GeneID 23248).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23248
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.