missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRD1B Antibody (4D6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00058490-M04
This item is not returnable.
View return policy
Description
RPRD1B Monoclonal antibody specifically detects RPRD1B in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| RPRD1B | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
| C20orf77 (AAH33629, 1 a.a. ∽ 326 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSSFSESALEKKLSEPSNSQHSVQTLSLWLIHHRKHAGPIVSVWHRELRKAKSNRKLTFLYLANDVIQNSKRKGPEFTREFESVLVDAFSHVAREADEGCKKPLERLLNIWQERSVYGGEFIQQLKLSMEDSKSPPPKATEEKKSLKRTFQQIQEEEDDDYPGSCSPQDPSAGPLLTEELIKALQDLENAASGDATVRQKIASLPQEVQDVSLLEKITDKEAAERLPKTVDEACLLLAEYNGRLAAELEDRRQLARMLVEYTQNQKDVLSEKEKKLEEYKQKLARVTQVRKELKSHIQSLPDLSLLPNVTGGLAPLPSAGDLFSTD | |
| 0.1 mg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 4D6 | |
| Western Blot 1:500, ELISA | |
| AAH33629 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 58490 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction