missing translation for 'onlineSavingsMsg'
Learn More

RPR-260243, MedChemExpress

Product Code. p-200101555
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487208

Brand: Medchem Express HY16915

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RPR-260243, a potent activator of human ether-a-go-go-related gene (hERG), slows deactivation and attenuates inactivation of hERG1 channels. RPR260243-modified HERG currents are inhibited by Dofetilide (IC50=58 nM). RPR260243 displays no activator-like effects on other voltage-dependent ion channels, including the closely related ERG3 K+ channel.

RPR-260243, a potent activator of human ether-a-go-go-related gene (hERG), slows deactivation and attenuates inactivation of hERG1 channels. RPR260243-modified HERG currents are inhibited by Dofetilide (IC50=58 nM). RPR260243 displays no activator-like effects on other voltage-dependent ion channels, including the closely related ERG3 K+ channel.

Specifications

Chemical Name or Material RPR-260243
CAS 668463-35-2
Color Light Brown
Purity Grade Notes Research
Molecular Formula C28H25F3N2O4
Quantity 10 mg
Solubility Information DMSO : 10 mg/mL (19.59 mM; Need ultrasonic)
SMILES O=C([C@H]1CN(CC#CC2=CC(F)=CC(F)=C2F)CC[C@H]1CCC(C3=CC=NC4=CC=C(OC)C=C34)=O)O
Molecular Weight (g/mol) 510.5
Formula Weight 510.5
Percent Purity 98.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.