missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPR-260243, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
RPR-260243, a potent activator of human ether-a-go-go-related gene (hERG), slows deactivation and attenuates inactivation of hERG1 channels. RPR260243-modified HERG currents are inhibited by Dofetilide (IC50=58 nM). RPR260243 displays no activator-like effects on other voltage-dependent ion channels, including the closely related ERG3 K+ channel.
Specifications
Specifications
| Chemical Name or Material | RPR-260243 |
| CAS | 668463-35-2 |
| Color | Light Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C28H25F3N2O4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 10 mg/mL (19.59 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1CN(CC#CC2=CC(F)=CC(F)=C2F)CC[C@H]1CCC(C3=CC=NC4=CC=C(OC)C=C34)=O)O |
| Molecular Weight (g/mol) | 510.5 |
| Formula Weight | 510.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction