missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPP40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595881
This item is not returnable.
View return policy
Description
Immunogen sequence: LSLNLDSKKY ERISWSFKEK KPLKFDFLLA WHKTGSEEST MMSYFSKYQI QEHQPKVALS TLRDLQCPVL QSSELEGTPE V.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends.
Specifications
| RPP40 | |
| Polyclonal | |
| Unconjugated | |
| RPP40 | |
| bA428J1.3; D8Bwg1265e; ribonuclease P (40 kD); ribonuclease P 40 subunit; ribonuclease P 40kDa subunit; ribonuclease P protein subunit p40; ribonuclease P, 40kD subunit; ribonuclease P/MRP 40 subunit; ribonuclease P/MRP 40kDa subunit; ribonuclease P/MRP subunit p40; ribonuclease P1; RNase P subunit 1; RNaseP protein p40; Rnasep1; RPP40 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10799, 208366, 291071 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.58 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75818, Q5BK64, Q8R1F9 | |
| RPP40 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-210 of human RPP40 (NP_001273061). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction