missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPP40 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93071-0.1ml
This item is not returnable.
View return policy
Description
RPP40 Polyclonal antibody specifically detects RPP40 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RPP40 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bA428J1.3,40kD subunit, EC 3.1.26.5, ribonuclease P/MRP 40kDa subunit, RNaseP protein p40 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-210 of human RPP40 (NP_001273061). LSLNLDSKKYERISWSFKEKKPLKFDFLLAWHKTGSEESTMMSYFSKYQIQEHQPKVALSTLRDLQCPVLQSSELEGTPEV | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10799 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction