missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPP25L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562567
This item is not returnable.
View return policy
Description
Immunogen sequence: LTKLRFLQTE DSWVPASPDT GLDPLTVRRH VPAVWVLLSR DPLDPNECGY QPPGAPPGLG SMPSSSCGPR SRRRARDTRS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
May be a component of ribonuclease P or MRP.
Specifications
| RPP25L | |
| Polyclonal | |
| Unconjugated | |
| RPP25L | |
| 2810432D09Rik; alba-like protein C9orf23; alba-like protein C9orf23 homolog; AW987258; bA296L22.5; C9orf23; RGD1306576; Ribonuclease P protein subunit p25-like protein; ribonuclease P/MRP 25 subunit-like; ribonuclease P/MRP 25kDa subunit-like; ribonuclease P/MRP subunit p25 like; RNase P protein subunit-like p25; Rpp25l; Rpp25-like protein; Rppl25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 138716 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N5L8 | |
| RPP25L | |
| Recombinant protein corresponding to Human RPP25L. Recombinant protein control fragment (Product #RP-93027). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction