Learn More
Invitrogen™ RPP25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597315
Description
Immunogen sequence: MENFRKVRSE EAPAGCGAEG GGPGSGPFAD LAPGAVHMRV KEGSKIRNLM AFATASMAQP ATRAIVFSGC GRATTKTVTC AEILKRRLAG LHQVTRLRYR SVREVWQSLP PGPTQGQTPG EPAASLSVLK NVPGLAILLS KDALDPRQPG YQPPNPHPGP SSPPAAPASK RSLGEPAAGE GSAKRSQPEP GVADEDQTA; Positive Samples: A-375, HeLa, HT-29; Cellular Location: Nucleus.
Component of ribonuclease P, a ribonucleoprotein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:12003489, PubMed:16723659, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt].
Specifications
| RPP25 | |
| Polyclonal | |
| Unconjugated | |
| RPP25 | |
| AI851155; ribonuclease P 25 subunit; ribonuclease P 25kDa subunit; ribonuclease P protein subunit p25; ribonuclease P/MRP 25 subunit; ribonuclease P/MRP 25kDa subunit; ribonuclease P/MRP subunit p25; RNase P protein subunit p25; Rpp25 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 102614, 54913 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91WE3, Q9BUL9 | |
| RPP25 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.